Assay Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110258.1 | NP_001103728.1 | 3 - 4 | 317 | 106 | |
GenBank mRNA | AF466349.1 | - | 3 - 4 | 317 | 106 | |
AY528236.1 | - | 3 - 4 | 291 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
