Assay Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110260.1 | NP_001103730.1 | 4 - 6 | 419 | 70 | |
GenBank mRNA | AF466352.1 | - | 4 - 5 | 419 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
