Assay Details
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032936.1 | NP_001028108.1 | 1 - 2 | 58 | 62 | |
GenBank mRNA | AY044445.1 | - | 1 - 2 | 58 | 62 | |
JU486844.1 | - | 1 - 2 | 89 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
