Assay Details
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032936.1 | NP_001028108.1 | 2 - 3 | 192 | 82 | |
XM_015138438.2 | XP_014993924.1 | 351 | 82 | |||
GenBank mRNA | AY044445.1 | - | 2 - 3 | 192 | 82 | |
JU486844.1 | - | 2 - 3 | 223 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
