Assay Details
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032936.1 | NP_001028108.1 | 3 - 4 | 274 | 148 | |
XM_015138438.2 | XP_014993924.1 | 433 | 148 | |||
GenBank mRNA | AY044445.1 | - | 3 - 4 | 274 | 148 | |
JU486844.1 | - | 3 - 4 | 305 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() |
