Assay Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032892.1 | NP_001028064.1 | 1 - 2 | 68 | 149 | |
GenBank mRNA | AY044446.1 | - | 1 - 2 | 68 | 149 | |
JU475477.1 | - | 1 - 2 | 117 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
