Assay Details
Gene Symbol: UTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032895.1 | NP_001028067.1 | 1 - 2 | 105 | 81 | |
GenBank mRNA | AY065983.1 | - | 1 - 2 | 105 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
