Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195417.1 | NP_001182346.1 | 4 - 5 | 457 | 63 | |
GenBank mRNA | AY369789.1 | - | 1 - 2 | 103 | 63 | |
CO726823.1 | - | 4 - 5 | 501 | 63 | ||
JU336288.1 | - | 4 - 5 | 588 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
