Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032910.1 | NP_001028082.1 | 4 - 5 | 1023 | 94 | |
GenBank mRNA | AY523632.1 | - | 4 - 5 | 1023 | 94 | |
AY523633.1 | - | 3 - 4 | 753 | 94 | ||
AY625001.1 | - | 3 - 4 | 753 | 94 | ||
DQ842021.1 | - | 3 - 4 | 753 | 94 | ||
EF113914.1 | - | 3 - 4 | 753 | 94 | ||
EF113915.1 | - | 3 - 4 | 753 | 94 | ||
EF113917.1 | - | 3 - 4 | 753 | 94 | ||
JU336825.1 | - | 4 - 5 | 874 | 94 | ||
JV048518.1 | - | 4 - 5 | 884 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
