Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032910.1 | NP_001028082.1 | 5 - 6 | 1038 | 96 | |
GenBank mRNA | AY523632.1 | - | 5 - 6 | 1038 | 96 | |
AY523633.1 | - | 4 - 5 | 768 | 96 | ||
AY625001.1 | - | 4 - 5 | 768 | 96 | ||
DQ842021.1 | - | 4 - 5 | 768 | 96 | ||
EF113914.1 | - | 4 - 5 | 768 | 96 | ||
EF113915.1 | - | 4 - 5 | 768 | 96 | ||
EF113917.1 | - | 4 - 5 | 768 | 96 | ||
JU336825.1 | - | 5 - 6 | 889 | 96 | ||
JV048518.1 | - | 5 - 6 | 899 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
