Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032910.1 | NP_001028082.1 | 7 - 8 | 1178 | 106 | |
GenBank mRNA | AY523632.1 | - | 7 - 8 | 1178 | 106 | |
AY625001.1 | - | 6 - 7 | 908 | 106 | ||
DQ842020.1 | - | 6 - 7 | 106 | |||
DQ842021.1 | - | 6 - 7 | 106 | |||
EF113914.1 | - | 6 - 7 | 908 | 106 | ||
EF113915.1 | - | 6 - 7 | 908 | 106 | ||
EF113916.1 | - | 6 - 7 | 106 | |||
EF113917.1 | - | 6 - 7 | 106 | |||
EF113918.1 | - | 6 - 7 | 106 | |||
JU336825.1 | - | 7 - 8 | 106 | |||
JU477573.1 | - | 7 - 8 | 106 | |||
JU477574.1 | - | 7 - 8 | 106 | |||
JU477575.1 | - | 7 - 8 | 106 | |||
JU477576.1 | - | 7 - 8 | 106 | |||
JU477577.1 | - | 7 - 8 | 106 | |||
JV048518.1 | - | 7 - 8 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
