Assay Details
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261638.1 | NP_001248567.1 | 6 - 7 | 588 | 138 | |
GenBank mRNA | DQ148142.1 | - | 2 - 3 | 195 | 138 | |
JU335752.1 | - | 6 - 7 | 570 | 138 | ||
JU471889.1 | - | 6 - 7 | 574 | 138 | ||
JV045701.1 | - | 6 - 7 | 588 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
