Assay Details
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035528.3 | NP_001030605.2 | 830 | 68 | ||
XM_028835460.1 | XP_028691293.1 | 1080 | 68 | |||
XM_028835461.1 | XP_028691294.1 | 819 | 68 | |||
GenBank mRNA | DQ188084.1 | - | 683 | 68 | ||
JU474298.1 | - | 786 | 68 | |||
JU474299.1 | - | 786 | 68 | |||
JU474300.1 | - | 786 | 68 | |||
JU474301.1 | - | 786 | 68 | |||
JU474302.1 | - | 786 | 68 | |||
JV046181.1 | - | 793 | 68 | |||
JV046182.1 | - | 793 | 68 | |||
JV601055.1 | - | 830 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
