Assay Details
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266489.1 | NP_001253418.1 | 210 | 73 | ||
GenBank mRNA | JU322430.1 | - | 210 | 73 | ||
JV704594.1 | - | 233 | 73 | |||
M96553.1 | - | 175 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
