Assay Details
Gene Symbol: SYPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266817.1 | NP_001253746.1 | 3 - 4 | 315 | 91 | |
GenBank mRNA | JU323179.1 | - | 3 - 4 | 317 | 91 | |
JU323242.1 | - | 2 - 3 | 299 | 91 | ||
JU476961.1 | - | 3 - 4 | 262 | 91 | ||
JV045068.1 | - | 3 - 4 | 315 | 91 | ||
JV703638.1 | - | 2 - 3 | 285 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
