Assay Details
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266993.1 | NP_001253922.1 | 416 | 103 | ||
XM_015135410.2 | XP_014990896.1 | 418 | 103 | |||
XM_015135411.2 | XP_014990897.1 | 536 | 103 | |||
XM_015135412.2 | XP_014990898.1 | 608 | 103 | |||
GenBank mRNA | JU472759.1 | - | 351 | 103 | ||
JV043268.1 | - | 416 | 103 | |||
JV653736.1 | - | 311 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
