Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193551.1 | NP_001180480.1 | 1 - 2 | 134 | 107 | |
GenBank mRNA | CN646333.1 | - | 1 - 2 | 134 | 107 | |
JU323241.1 | - | 1 - 2 | 168 | 107 | ||
JU475742.1 | - | 1 - 2 | 88 | 107 | ||
JV045361.1 | - | 1 - 2 | 87 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193551.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
