Assay Details
Gene Symbol: POLR2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145794.2 | XP_015001280.1 | 673 | 83 | ||
XM_015145795.2 | XP_015001281.1 | 288 | 83 | |||
XM_015145796.2 | XP_015001282.1 | 303 | 83 | |||
XM_028852798.1 | XP_028708631.1 | 290 | 83 | |||
GenBank mRNA | JU473271.1 | - | 220 | 83 | ||
JV567594.1 | - | 233 | 83 | |||
JV639047.1 | - | 262 | 83 |
Gene Symbol: LOC100426798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129076.2 | XP_014984562.1 | 184 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
