Assay Details
Gene Symbol: MRPL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193419.1 | NP_001180348.1 | 51 | 71 | ||
GenBank mRNA | CN804367.1 | - | 51 | 71 | ||
JV490915.1 | - | 96 | 71 | |||
JV601282.1 | - | 81 | 71 |
Gene Symbol: LOC100430397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002799391.3 | XP_002799437.2 | 89 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
