Assay Details
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083632.4 | XP_001083632.1 | 408 | 92 | ||
XM_001084303.4 | XP_001084303.1 | 788 | 92 | |||
XM_002804108.3 | XP_002804154.1 | 906 | 92 | |||
XM_015138629.2 | XP_014994115.1 | 785 | 92 | |||
XM_015138630.2 | XP_014994116.1 | 410 | 92 | |||
XM_015138631.2 | XP_014994117.1 | 452 | 92 | |||
XM_015138632.2 | XP_014994118.1 | 449 | 92 | |||
XM_015138635.2 | XP_014994121.1 | 315 | 92 | |||
XM_028848557.1 | XP_028704390.1 | 903 | 92 | |||
XM_028848558.1 | XP_028704391.1 | 314 | 92 | |||
GenBank mRNA | JU349577.1 | - | 2 - 3 | 91 | ||
JV489647.1 | - | 2 - 3 | 594 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
