Assay Details
Gene Symbol: MEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194869.1 | NP_001181798.1 | 2 - 3 | 399 | 97 | |
XM_001087765.4 | XP_001087765.2 | 422 | 97 |
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335298.1 | - | 17 - 18 | 3333 | 97 | |
JU471102.1 | - | 17 - 18 | 3329 | 97 | ||
JV045007.1 | - | 17 - 18 | 3332 | 97 | ||
JV566389.1 | - | 17 - 18 | 3341 | 97 | ||
JV600899.1 | - | 17 - 18 | 3294 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
