Assay Details
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260748.1 | NP_001247677.1 | 587 | 80 | ||
XM_028852120.1 | XP_028707953.1 | 531 | 80 | |||
GenBank mRNA | JU335246.1 | - | 587 | 80 | ||
JV586068.1 | - | 572 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
