Assay Details
Gene Symbol: AIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138918.2 | XP_014994404.1 | 385 | 145 | ||
GenBank mRNA | JU350608.1 | - | 3 - 4 | 318 | 145 | |
JV487119.1 | - | 3 - 4 | 338 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
