Assay Details
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266004.2 | NP_001252933.1 | 470 | 57 | ||
GenBank mRNA | JU320545.1 | - | 360 | 57 | ||
JU475208.1 | - | 235 | 57 | |||
JV043273.1 | - | 470 | 57 | |||
JV669286.1 | - | 385 | 57 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
