Assay Details
Gene Symbol: SAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085118.3 | XP_001085118.2 | 1320 | 83 | ||
XM_015122572.2 | XP_014978058.1 | 1315 | 83 | |||
XM_015122574.2 | XP_014978060.1 | 1320 | 83 | |||
XM_015122575.2 | XP_014978061.1 | 1320 | 83 | |||
XM_028839739.1 | XP_028695572.1 | 1016 | 83 | |||
XM_028839740.1 | XP_028695573.1 | 883 | 83 | |||
GenBank mRNA | JU335835.1 | - | 1221 | 83 | ||
JU335836.1 | - | 1203 | 83 | |||
JU335837.1 | - | 1221 | 83 | |||
JU476714.1 | - | 1153 | 83 | |||
JU476717.1 | - | 1153 | 83 | |||
JV047883.1 | - | 1251 | 83 | |||
JV047884.1 | - | 1201 | 83 | |||
JV554113.1 | - | 1207 | 83 | |||
JV600312.1 | - | 1184 | 83 |
Gene Symbol: SAFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU337263.1 | - | 1221 | 83 | ||
JU476715.1 | - | 1153 | 83 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
