Assay Details
Gene Symbol: CSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257285.1 | NP_001244214.1 | 464 | 64 | ||
XM_015118823.2 | XP_014974309.2 | 938 | 64 | |||
XM_015118831.2 | XP_014974317.1 | 529 | 64 | |||
XM_028843664.1 | XP_028699497.1 | 589 | 64 | |||
GenBank mRNA | BQ807632.1 | - | 464 | 64 | ||
BQ807767.1 | - | 288 | 64 | |||
JU474399.1 | - | 494 | 64 | |||
JV673835.1 | - | 484 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
