Assay Details
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266219.2 | NP_001253148.1 | 228 | 95 | ||
GenBank mRNA | JU320282.1 | - | 240 | 95 | ||
JU470709.1 | - | 235 | 95 | |||
JV044801.1 | - | 231 | 95 | |||
JV675335.1 | - | 217 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
