Assay Details
Gene Symbol: UBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261050.1 | NP_001247979.1 | 3 - 4 | 258 | 65 | |
XM_015122785.2 | XP_014978271.1 | 296 | 65 | |||
XM_028838446.1 | XP_028694279.1 | 423 | 65 | |||
GenBank mRNA | JU471006.1 | - | 3 - 4 | 258 | 65 | |
JV601589.1 | - | 3 - 4 | 232 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
