Assay Details
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258140.1 | NP_001245069.1 | 1 - 2 | 146 | 77 | |
GenBank mRNA | JU473371.1 | - | 1 - 2 | 146 | 77 | |
JV045700.1 | - | 1 - 2 | 100 | 77 | ||
JV635877.1 | - | 1 - 2 | 135 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001258140.1 transcripts.
This assay is in the 5'UTR region of the NM_001258140.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
