Assay Details
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258140.1 | NP_001245069.1 | 2 - 3 | 270 | 126 | |
GenBank mRNA | JU473371.1 | - | 2 - 3 | 270 | 126 | |
JV045700.1 | - | 2 - 3 | 224 | 126 | ||
JV635877.1 | - | 2 - 3 | 259 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
