Assay Details
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168614.1 | NP_001162085.1 | 1 - 2 | 96 | 91 | |
GenBank mRNA | CO579359.1 | - | 1 - 2 | 96 | 91 | |
JU336862.1 | - | 1 - 2 | 115 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001168614.1 transcripts.
This assay is in the 5'UTR region of the NM_001168614.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
