Assay Details
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168614.1 | NP_001162085.1 | 251 | 68 | ||
XM_015134736.2 | XP_014990222.1 | 874 | 68 | |||
XM_028844899.1 | XP_028700732.1 | 263 | 68 | |||
GenBank mRNA | CO579359.1 | - | 251 | 68 | ||
JU336862.1 | - | 270 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
