Assay Details
Gene Symbol: C1QC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130170.2 | XP_014985656.1 | 129 | 59 | ||
GenBank mRNA | JU323143.1 | - | 72 | 59 | ||
JV490269.1 | - | 91 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015130170.2 transcripts.
Set Membership: |
![]() ![]() |
