Assay Details
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266350.2 | NP_001253279.1 | 466 | 137 | ||
GenBank mRNA | JU319664.1 | - | 446 | 137 | ||
JU473005.1 | - | 408 | 137 | |||
JV044789.1 | - | 468 | 137 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
