Assay Details
Gene Symbol: RPS19BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099808.4 | XP_001099808.2 | 394 | 50 | ||
GenBank mRNA | JV700997.1 | - | 3 - 4 | 356 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
