Assay Details
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194815.3 | NP_001181744.2 | 410 | 85 | ||
GenBank mRNA | JU477220.1 | - | 395 | 85 | ||
JV048260.1 | - | 410 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
