Assay Details
Gene Symbol: WDR83OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194406.1 | NP_001181335.1 | 305 | 60 | ||
GenBank mRNA | JU320875.1 | - | 214 | 60 | ||
JU470489.1 | - | 149 | 60 | |||
JV043684.1 | - | 89 | 60 | |||
JV566224.1 | - | 306 | 60 | |||
JV639485.1 | - | 258 | 60 |

More Information
Set Membership: |
![]() ![]() |
