Assay Details
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194897.3 | NP_001181826.2 | 1 - 2 | 207 | 87 | |
GenBank mRNA | JU322906.1 | - | 1 - 2 | 200 | 87 | |
JU475861.1 | - | 1 - 2 | 191 | 87 | ||
JV686060.1 | - | 1 - 2 | 207 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
