Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193491.1 | NP_001180420.1 | 5 - 6 | 737 | 68 | |
XM_015123082.2 | XP_014978568.1 | 1081 | 68 | |||
XM_015123083.2 | XP_014978569.1 | 1895 | 68 | |||
XM_015123084.1 | XP_014978570.1 | 880 | 68 | |||
XM_015123085.2 | XP_014978571.1 | 860 | 68 | |||
GenBank mRNA | JU327645.1 | - | 5 - 6 | 579 | 68 | |
JU473916.1 | - | 6 - 7 | 836 | 68 | ||
JV045867.1 | - | 6 - 7 | 800 | 68 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
