Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113359.1 | NP_001106830.1 | 4 - 5 | 775 | 69 | |
XM_015115035.2 | XP_014970521.1 | 1088 | 69 | |||
GenBank mRNA | EU124697.1 | - | 4 - 5 | 775 | 69 | |
JU332480.1 | - | 5 - 6 | 920 | 69 | ||
JU471017.1 | - | 5 - 6 | 926 | 69 | ||
JV045892.1 | - | 5 - 6 | 938 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
