Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113359.1 | NP_001106830.1 | 5 - 6 | 870 | 106 | |
XM_015115035.2 | XP_014970521.1 | 1183 | 106 | |||
GenBank mRNA | EU124697.1 | - | 5 - 6 | 870 | 106 | |
JU332480.1 | - | 6 - 7 | 1015 | 106 | ||
JU471017.1 | - | 6 - 7 | 1021 | 106 | ||
JV045892.1 | - | 6 - 7 | 1033 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
