Assay Details
Gene Symbol: MRPL50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194797.3 | NP_001181726.2 | 1 - 2 | 96 | 63 | |
XM_001111283.4 | XP_001111283.2 | 186 | 63 | |||
GenBank mRNA | JU335090.1 | - | 1 - 2 | 103 | 63 | |
JV047438.1 | - | 1 - 2 | 103 | 63 | ||
JV654243.1 | - | 1 - 2 | 129 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
