Assay Details
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194595.2 | NP_001181524.2 | 5 - 6 | 714 | 64 | |
GenBank mRNA | JU325226.1 | - | 4 - 5 | 548 | 64 | |
JV481845.1 | - | 5 - 6 | 667 | 64 | ||
JV593627.1 | - | 5 - 6 | 657 | 64 | ||
JV628728.1 | - | 5 - 6 | 650 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
