Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107808.4 | XP_001107808.1 | 584 | 145 | ||
XM_001107873.3 | XP_001107873.1 | 530 | 145 | |||
XM_015152780.2 | XP_015008266.1 | 520 | 145 | |||
GenBank mRNA | JU328140.1 | - | 301 | 145 | ||
JV547659.1 | - | 358 | 145 | |||
JV686659.1 | - | 421 | 145 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
