Assay Details
Gene Symbol: PRXL2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106503.4 | XP_001106503.1 | 580 | 124 | ||
GenBank mRNA | JU326556.1 | - | 5 - 6 | 493 | 124 | |
JV708795.1 | - | 5 - 6 | 503 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
