Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109452.4 | XP_001109452.2 | 436 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109452.4 transcripts.
Set Membership: |
![]() ![]() |
