Assay Details
Gene Symbol: KRBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108465.4 | XP_001108465.2 | 107 | 128 | ||
XM_015131215.2 | XP_014986701.1 | 107 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001108465.4, XM_015131215.2 transcripts.
Set Membership: |
![]() ![]() |
