Assay Details
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106814.4 | XP_001106814.3 | 373 | 63 | ||
XM_015118134.2 | XP_014973620.2 | 327 | 63 | |||
XM_028835374.1 | XP_028691207.1 | 261 | 63 | |||
XM_028835375.1 | XP_028691208.1 | 267 | 63 | |||
XM_028835376.1 | XP_028691209.1 | 325 | 63 | |||
XM_028835377.1 | XP_028691210.1 | 274 | 63 | |||
GenBank mRNA | JU323374.1 | - | 187 | 63 | ||
JU481076.1 | - | 124 | 63 | |||
JV044398.1 | - | 160 | 63 | |||
JV637693.1 | - | 187 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
