Assay Details
Gene Symbol: EIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257885.1 | NP_001244814.1 | 450 | 80 | ||
GenBank mRNA | JU320521.1 | - | 450 | 80 | ||
JV567412.1 | - | 556 | 80 | |||
JV602600.1 | - | 554 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
