Assay Details
Gene Symbol: RPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265719.1 | NP_001252648.1 | 349 | 117 | ||
XM_028838449.1 | XP_028694282.1 | 451 | 117 | |||
GenBank mRNA | JV528877.1 | - | 389 | 117 | ||
JV565041.1 | - | 349 | 117 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
